Polysorbate 20 (vegetable based) NF, Multi-Compendial, refined, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:9005-64-5
:
Formula:C₅₈H₁₁₄O₂₆
MDL Number:MFCD00165986
Density:1.08 kg (1 L)
Flash Pt:148.00 °C
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Ethylene glycol:≤ 0.10 %
- Diethylene glycol:≤ 0.10 %
- GMP Manufactured Product:≤2.0
- Meets E.P. Chemical Specifications:≤0.25%
- Meets B.P. Chemical Specifications:≤10 ppm
- Meets JPE Specifications:≤1 ppm
- Meets N.F. Requirements:≤10 ppm
- CAUTION: For Manufacturing, processing or repackaging:96 - 108
- Bulk Pharmaceutical Chemical:Passes Test
- NF - Acid Value:≤2.0
- NF - Residue on Ignition:≤0.25 %
- NF - Dioxane:≤10 ppm
- NF - Ethylene Oxide:≤1 ppm
- NF - Hydroxyl Value, mg KOH/g:96 - 108
- NF - Identification A:Passes Test
- NF - Identification B:Passes Test
- NF - Saponification Value, mg KOH/g:40 - 50
- NF - Caproic Acid (Composition of Fatty Acids):≤1.0 %
- NF - Caprylic Acid (Composition of Fatty Acids):≤10.0 %
- NF - Capric Acid (Composition of Fatty Acids):≤10.0 %
- NF - Lauric Acid (Composition of Fatty Acids):40.0 - 60.0 %
- NF - Myristic Acid (Composition of Fatty Acids):14.0 - 25.0 %
- NF - Palmitic Acid (Composition of Fatty Acids):7.0 - 15.0 %
- NF - Stearic Acid (Composition of Fatty Acids):≤11.0 %
- NF - Oleic Acid (Composition of Fatty Acids):≤11.0 %
- NF - Linoleic Acid (Composition of Fatty Acids):≤3.0 %
- NF - Peroxide Value:≤5.0
- NF - Water (H₂O):≤3.0 %
- EP/BP - Acid Value:≤2.0
- EP/BP - Total Ash:≤0.25 %
- EP/BP - Hydroxyl Value:96 - 108
- EP/BP - Identification A:Passes Test
- EP/BP - Identification D:Passes Test
- EP/BP - Saponification Value:40 - 50
- EP/BP - Ethylene Oxide:≤1 ppm
- EP/BP - Dioxan:≤10 ppm
- EP/BP - Water (H₂O):≤3.0 %
- EP/BP - Composition of Fatty Acids - Caproic Acid:≤1.0 %
- EP/BP - Composition of Fatty Acids - Caprylic Acid:≤10.0 %
- EP/BP - Composition of Fatty Acids - Capric Acid:≤10.0 %
- EP/BP - Composition of Fatty Acids - Lauric Acid:40.0 - 60.0
- EP/BP - Composition of Fatty Acids - Myristic Acid:14.0 - 25.0 %
- EP/BP - Composition of Fatty Acids - Palmitic Acid:7.0 - 15.0
- EP/BP - Composition of Fatty Acids - Stearic Acid:≤7.0 %
- EP/BP - Composition of Fatty Acids - Oleic Acid:≤11.0 %
- EP/BP - Composition of Fatty Acids - Linoleic Acid:≤3.0 %
- EP - Peroxide Value:≤10.0
- JPE - Identification 1:Passes Test
- JPE - Identification 2:Passes Test
- JPE - Identification 3:Passes Test
- JPE - pH at 25°C (5% Aqueous):4.0 - 7.0
- JPE - Water (H₂O):≤3.0 %
- JPE - Acid Value:≤4.0
- JPE - Saponification Value:43 - 55
- JPE - pH:4.0 - 7.0
- JPE - Residue on Ignition:≤0.25 %
- JPE - Specific Gravity at 20°C:1.090 - 1.130
- JPE - Viscosity (mm²/S):350 - 550
- JPE - Heavy Metals (as Pb):≤20 ppm
- JPE - Arsenic (As):≤2 ppm
- Microbiological - Total Aerobic Microbial Count (cfu/g):≤100
- Microbiological - Escherichia coli:Passes Test
- Microbiological - Pseudomonas aeruginosa:Passes Test
- Microbiological - Salmonella:Passes Test
- Microbiological - Staphylococcus aureus:Passes Test
- Microbiological - Total Yeast and Mold Count (cfu/g):≤50
- Endotoxin Concentration (EU/g):≤10
- Color (APHA):≤150
- Formaldehyde:≤10 ppm
- Potassium (K):≤5 ppm
- Sodium (Na):≤5 ppm