Order Entry
China
ContactUsLinkComponent
Magnesium acetate tetrahydrate 98.0-102.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Magnesium acetate tetrahydrate 98.0-102.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # BAKR2425-07
Supplier:  Avantor
CAS Number:  
undefined
Magnesium acetate tetrahydrate 98.0-102.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # BAKR2425-07
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    PE pail, screw top
  • Pk:
    12 kg
  • Seq:
    4187

Specifications

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
  • For Laboratory,Research,or Manufacturing Use:
  • ACS - Assay ((CH₃COO)₂Mg·4H₂O):
    98.0 - 102.0 %
  • ACS - Insoluble Matter:
    ≤0.005 %
  • ACS - pH of 5% Solution at 25°C:
    6.5 - 8.5
  • ACS - Chloride (Cl):
    ≤0.001 %
  • ACS - Sulfate (SO₄):
    ≤0.005 %
  • ACS - Barium (Ba):
    ≤0.001 %
  • ACS - Calcium (Ca):
    ≤0.01 %
  • ACS - Manganese (Mn):
    ≤0.001 %
  • ACS - Potassium (K):
    ≤0.005 %
  • ACS - Sodium (Na):
    ≤0.005 %
  • ACS - Strontium (Sr):
    ≤0.005 %
  • ACS - Endotoxin Concentration (2.5 EU/g max.):
    Passes Test
  • Trace Impurities - ACS - Heavy Metals (as Pb):
    ≤5 ppm
  • Trace Impurities - Iron (Fe):
    ≤5 ppm

Specification Test Results

About this item

 :
 :214.46 g/mol
Density:1.45 g/cm³
CAS Number:16674-78-5
Formula:(H₃CCOO)₂Mg·4H₂O
MDL Number:MFCD00149214