


Ammonia solution 28.0 - 30.0% (NH₃ basis), AR® ACS (26° Baume), Macron Fine Chemicals™
:1336-21-6
:7664-41-7
:NH₃
:MFCD00066650
:35.05 g/mol
:32…37.7 °C (1013 hPa)
:–60…–57.5 °C
:0.90 g/cm³
:215-647-6
:2672
:8,III
:GHS05
:GHS07
:GHS09
:Ambient
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Meets ACS Reagent Chemical Requirements:28.0 - 30.0%
- For Laboratory,Research,or Manufacturing Use:≤0.002%
- ACS - Assay (as NH₃):28.0 - 30.0 %
- Appearance:Passes Test
- ACS - Carbon Dioxide (CO₂):≤ 0.002 %
- ACS - Residue after Ignition:≤ 0.002 %
- ACS - Substances Reducing Permanganate:Passes Test
- Arsenic and Antimony (as As):≤ 3 ppm
- Chloride (Cl):≤ 0.5 ppm
- Trace Impurities - Heavy Metals (as Pb):≤ 0.5 ppm
- Nitrate (NO₃):≤ 2 ppm
- Phosphate (PO₄):≤ 2 ppm
- Sulfate (SO₄):≤ 2 ppm
- Trace Impurities - Aluminum (Al):≤ 0.300 ppm
- Trace Impurities - Boron (B):≤ 0.100 ppm
- Trace Impurities - Calcium (Ca):≤ 0.300 ppm
- Trace Impurities - Chromium (Cr):≤ 0.200 ppm
- Trace Impurities - Copper (Cu):≤ 0.10 ppm
- Trace Impurities - Gold (Au):≤ 0.300 ppm
- Trace Impurities - Iron (Fe):≤ 0.2 ppm
- Trace Impurities - Lead (Pb):≤ 0.200 ppm
- Trace Impurities - Magnesium (Mg):≤ 0.300 ppm
- Trace Impurities - Manganese (Mn):≤ 0.200 ppm
- Trace Impurities - Nickel (Ni):≤ 0.1 ppm
- Trace Impurities - Potassium (K):≤ 0.300 ppm
- Trace Impurities - Sodium (Na):≤ 2.000 ppm
- Trace Impurities - Tin (Sn):≤ 0.300 ppm
- Trace Impurities - Titanium (Ti):≤ 0.300 ppm
- Trace Impurities - Zinc (Zn):≤ 0.300 ppm