
Ammonium chloride ≥99.5% (by argentometric titration), crystals, AR® ACS, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Certificates
About this item
CAS Number:12125-02-9
:MFCD00143604
Formula:NH₄Cl
MDL Number:MFCD00011420
:53.49 g/mol
Merck Index:13,00510
Melting Pt:335 °C
Density:1.53 g/cm³
:Management of Change (MOC) category = N
EINECS:235-186-4
:GHS07
Storage Temperature:Ambient
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Meets ACS Reagent Chemical Requirements:≥99.5%
- For Laboratory,Research,or Manufacturing Use:≤0.001%
- ACS - Assay (NH₄Cl) (by Ag titrn):≥99.5 %
- ACS - Calcium (Ca):≤0.001 %
- ACS - Insoluble Matter:≤0.005 %
- ACS - Iron (Fe):≤2 ppm
- ACS - Magnesium (Mg):≤5 ppm
- ACS - pH of 5% Solution at 25°C:4.5 - 5.5
- ACS - Phosphate (PO₄):≤2 ppm
- ACS - Residue after Ignition:≤0.01 %
- ACS - Sulfate (SO₄):≤0.002 %
- ACS - Appearance (white crystals):Passes Test
- Heavy Metals (as Pb):≤2 ppm
- Nitrate (NO₃):≤5 ppm