Order Entry
China
ContactUsLinkComponent
Luciferase assay system, Bio-Glo™
Luciferase assay system, Bio-Glo™
Supplier:  PROMEGA
Certificates
undefined
Luciferase assay system, Bio-Glo™
Supplier:  PROMEGA

Specifications

  • Storage conditions:
    Store the Bio-Glo Luciferase Assay System components at –30...–10 °C. The Bio-Glo Luciferase Assay Buffer can be stored at below 30 °C for up to three months with approximately a 10% change in reagent functionality. For optimal performance, reconstituted Bio-Glo Luciferase Assay Reagent should be used the day of preparation. However, once reconstituted, Bio-Glo Luciferase Assay Reagent can be stored at –20 °C for up to six weeks.
  • Storage temperature:
    Room temperature or 4 °C storage
  • Application:
    For use in the ADCC reporter bioassay; Reporter-based bioassays

Specifications