Dual-Luciferase® reporter (DLR™) assay system
Catalog # PROME1960
Supplier: PROMEGA
CAS Number:
Specifications
- Pk:1.000 Assays
- Description:Dual-Luciferase® Reporter Assay, 10×100 assays
- Seq:02
Specifications
About this item
The Dual-Luciferase® Reporter (DLR™) Assay System provides an efficient means of performing two reporter assays. In the DLR™ Assay, the activities of firefly (Photinus pyralis) and Renilla (Renilla reniformis or sea pansy) luciferases are measured sequentially from a single sample.
- Renilla luciferase internal control provides more accurate results
- Range extends over 7 logs; very active samples typically do not need dilution
- Requires luminometer with 2 injectors
The pGL4 Luciferase Reporter Vectors are designed for use with the DLR™ Assay Systems. A Renilla luciferase vector with constitutive expression may be used in combination with any experimental firefly luciferase vector to co-transfect mammalian cells.
Delivery: For applications requiring more lysis reagent (e.g., >100 µl/well), additional Passive Lysis Buffer may be purchased separately.