Avantor® Apollo C8, HPLC Columns
Supplier: HICHROM
Certificates
About this item
Avantor® Apollo HPLC columns use high purity, base deactivated silica for powerful separations.
- Easy scale-up from analytical to preparative
- Extended pH stability is about 1,5 to 10,5
- USP L7
These columns are ideal for routine analysis and for educational laboratories.
Specifications
- Catalog No:
- HICH36512
- HICH5126774
- HICH36506
- HICH96463/N
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)