Order Entry
China
ContactUsLinkComponent
Jars, cloud and pour point, PYREX®
  CORN6902
undefined
Jars, cloud and pour point, PYREX®
  CORN6902
  • Ext.Ø:
    35 mm
  • Height:
    121 mm
  • Seq:
    01
  • Pk:
    1
Made of PYREX® glass, clear.
  • Not intended for vacuum use
  • Good chemical resistance
  • Good temperature resistance
These jars are used to measure the cloud point temperature in petroleum oils. A single line encircles the jar at a point approximately 51 mm above the bottom to assist in measuring the sample.
 : Meets ASTM D-97, D-1500, D-2500 specifications.