HPLC columns, YMC-Pack™ YMC Carotenoid (C30)
Catalog # YMCACT99S032546
Supplier: YMC
Specifications
- Column Format:Analytical
- Ø int.:4,60 mm
- Particle Size:3 µm
- Length:250 mm
- Phase:C30
- Pk:1
Specifications
About this item
The YMC30 is a very lipophilic phase with exceptional selectivity towards isomeric structures. The C30 ligands provide sufficient phase thickness to enhance interaction with long chain molecules. Particularly suited for the separation of carotenoids.
- More hydrophobic
- Provides sufficient phase thickness to enhance interaction with long chained molecules
- Geometric and positional isomers of conjugated double bonding systems can be recognised and resolved
Ideal for the separation of polar carotenes, polar and nonpolar xanthophylls, steroids, retinols, fat-soluble vitamins and LC-MS applications. Also used in the food industry, for the analysis of vitamin formulations, in environmental analysis, and for the control of algal growth. Other potential applications include the separation of prostaglandins and leucotrienes.
Delivery: Every column supplied with lot certificate on physicochemical properties of bare silica, minimal metal traces, bonding and chromatographic performance; test chromatogram showing high performance plate count together with tight specification on peak symmetry.