Order Entry
China
ContactUsLinkComponent
HPLC columns, YMC-Pack™ ODS-AQ
HPLC columns, YMC-Pack™ ODS-AQ
Supplier:  YMC
Certificates
HPLC columns, YMC-Pack™ ODS-AQ
Supplier:  YMC
Specifications
  • Catalog No:
  • YMCAAQ12S03-0546WT
  • YMCAAQ12S05-2546WT
  • Carbon load:
  • 14%
  • 14%
  • Endcapped:
  • Yes
  • Yes
  • pH range:
  • 2,0 - 7,5
  • 2,0 - 7,5
  • Surface area:
  • 330 m²/g
  • 330 m²/g
  • USP code:
  • L1
  • L1
Specifications