Order Entry
China
ContactUsLinkComponent
pH/mV/°C meters, handheld, SensION+ MM110
pH/mV/°C meters, handheld, SensION+ MM110
  HACHLPV2645970002
 :  HACH
undefined
pH/mV/°C meters, handheld, SensION+ MM110
  HACHLPV2645970002
 :  HACH

 

  • Temperature range:
    −20...+150 °C
  • Description:
    Handheld multi-parameter meter kit for pH, ORP and temperature, MM110
  • Seq:
    01
  • Model:
    MM110
  • Temperature accuracy:
    ±0,2 °C
  • Temperature resolution:
    0,1 °C
  • Automatic buffer recognition:
    pH 2,01/4,01/7,00/9,21/10,05
  • Connections:
    MP-8
  • Data logging capacity:
    500 results
  • Display:
    Backlit LCD with pictograms, 128×64 pixel
  • Languages:
    Icon based, no language needed
  • Outputs:
    Wireless to USB
  • Power supply:
    3 AA batteries
  • Temperature compensation:
    With temperature probe Pt 1000 or by keypad
  • Weight:
    0,3 kg
  • W×D×H:
    73×186×38 mm
  • mV accuracy:
    ±1
  • mV range:
    ±1999
  • mV resolution:
    1
  • pH accuracy:
    ≤ 0,02
  • pH calibration:
    1 - 3 point
  • pH range:
    −2...19,99
  • pH resolution:
    0,01
  • Pk:
    1

 

 

These compact meters are capable of measuring 3 (pH, ORP and temperature) parameters simultaneously. The meters are designed for field use.

  • A single probe provides pH, ORP and temperature simultaneously
  • Maximum simplicity: sensION+ systems come complete with everything you need to start testing
  • sensION+ carrying cases work as complete, portable labs
  • Screw-on calibration tubes and probe storage tubes simplify use in field
  • IP67-protected

MM110 is ideal for for Pool and Spa and general aqueous samples.

 : Supplied with sensION+ pH1 meter, sensION+ 5045 multi sensor for pH and ORP, sensION+ portable field carrying case, 125 ml of pH 4, 7 and 10 calibration standards, 125 ml of ORP calibration standard 220 mV, colour coded screwable calibration tubes,125 ml ORP calibration standard, sample and measurement containers, and user documentation.