Hydrogen peroxide test kit, CHEMets®
Catalog # CHETR-5510
Supplier: CHEMETRICS
CAS Number:
Specifications
- Chemical method:Ferric thiocyanate
- Pk:1 KIT
- Description:CHEMets refill pack, 30 ampoules
- Seq:02
- Range:0 - 0,8; 1 - 10 ppm
Specifications
About this item
Hydrogen Peroxide CHEMets® used to to monitor sterilisation solutions in the packaging and sanitising processes. Hydrogen peroxide is a strong oxidising agent used for treating of industrial effluents and domestic waste and serving as a disinfectant in aseptic packaging.
Delivery: Test kits contain 30 ampules refill packs, comparator(s), accessory solutions (when necessary), 25 ml sample cup, and instructions.