Order Entry
China
ContactUsLinkComponent
Non-ionic detergent, Kolliphor® HS 15
Non-ionic detergent, Kolliphor® HS 15
Catalog # SIAL42966-1KG
Supplier:  SIGMAALDRICH
Non-ionic detergent, Kolliphor® HS 15
Catalog # SIAL42966-1KG
Supplier:  SIGMAALDRICH
Specifications
  • Description:
    Kolliphor® HS 15
  • Pk:
    1 kg
  • pH range:
    5,0 - 7,0
Specifications
About this item
Kolliphor HS 15 is a non-ionic detergent obtained by reacting 15 moles of ethylene oxide with 1 mole of 12-hydroxy stearic acid. It's a non-ionic solubiliser and emulsifier.