Luciferase assay system, Steady-Glo®
Supplier: PROMEGA
Certificates
About this item
High-throughput quantitation of firefly (Photinus pyralis) luciferase expression in mammalian cells is commonly performed by batch processing of 96- and 384-well plates. Steady-Glo® Luciferase Assay System is designed for this purpose of providing long-lived luminescence when added to cultured cells. The homogeneous assay provides signal half-lives of over 5 hours in commonly used cell culture media, and does not require sample preprocessing. Throughput rates of several thousand samples per hour may be achieved with high reproducibility under standard laboratory conditions.
- Stable luminescence signal with 5-hour half-life
- No need to remove culture medium or wash cells prior to adding assay reagent
- Less sensitive to mixing conditions in multiwell plates, particularly useful in 384-well plates