Order Entry
China
Orders LinkContactUsLinkComponent
Vectashield® mounting medium
Vectashield® mounting medium, 10 mL
  VECTH-1000
Vectashield® mounting medium
  VECTH-1000
 :  
Vectashield® mounting medium
Vectashield® mounting medium, 10 mL
 :  
1 * 10 毫升
Vectashield® Mounting Medium is a unique, stable formula for preserving fluorescence. It is superior to other anti-fading reagents or mounting media for fluorescence microscopy. Some agents retard the rate of fading, but their initial application gives a severe reduction in emission. Others give a good initial reading but bleach upon illumination. Vectashield® Mounting Medium combines both advantages into one product. It provides strong initial fluorescence and retards photobleaching over time. Vectashield® Mounting Medium is supplied in a bottle fitted with a cone-seal cap. A cap fitted with a drop dispenser pipette is also included. The cone-seal cap can be replaced with the drop dispenser cap for routine use and during storage. Mounted slides should be stored at 4 °C. The fluorescence will remain for many weeks.
  • Prevents rapid loss of fluorescence during microscopic examination
  • Retains its anti-fading ability during long-term storage
  • Optically clear
  • Mounting medium to preserve fluorescence and counterstain DNA with DAPI (blue)
  • Pk:
    10 mL
  • Description:
    Vectashield® mounting medium