Order Entry
China
ContactUsLinkComponent
Adapters, expansion, cone on socket
  CGLACG-1002-07
 :  CHEMGLASS
undefined
Adapters, expansion, cone on socket
  CGLACG-1002-07
 :  CHEMGLASS
  • Cone size:
    24/40 NS
  • Socket size:
    29/42 NS
  • Pk:
    1
Used to connect dissimilar standard taper joint sizes. Expansion adapter from small cone to large socket.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes