Order Entry
China
ContactUsLinkComponent
Staphylococcus aureus (Strain V8) protease V8, MP Biomedicals
Staphylococcus aureus (Strain V8) protease V8, MP Biomedicals
Catalog # ICNA08399001
Supplier:  MP BIOMEDICALS
CAS Number:  
Staphylococcus aureus (Strain V8) protease V8, MP Biomedicals
Catalog # ICNA08399001
Supplier:  MP BIOMEDICALS
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pk:
    5 mg
  • Species:
    Staphylococcus aureus (Strain V8)
  • Storage conditions:
    2...8 °C
  • Form:
    Lyophilized powder
  • Enzyme name:
    Protease V8
  • Enzyme activity:
    915 units/mg (dry weight)
  • Molecular weight:
    27,7 kDa
  • pH:
    4,0/7,8

Specifications

About this item

Protease V8 specifically cleaves peptide bonds on the COOH terminal side of either aspartic or glutamic acids. In the presence of ammonium, the specificity is limited to glutamic sites.

It is used in a study to assess enzymatic cleaning of inorganic ultrafiltration membranes used for whey protein fractionation. It has also been used in a study to investigate immobilised residue-specific endoproteinases for protein sequencing.

Unit definition: One unit is that amount of enzyme required to cause a change of 0,001 A280 nm per minute at 37 °C, pH 7,8, using casein as the substrate.