Order Entry
China
ContactUsLinkComponent
Flasks, round bottom, glass, clear, standard ground joints, two necks
Flasks, round bottom, glass, clear, standard ground joints, two necks
Catalog # CGLACG-1520-02
Supplier:  CHEMGLASS
undefined
Flasks, round bottom, glass, clear, standard ground joints, two necks
Catalog # CGLACG-1520-02
Supplier:  CHEMGLASS
Specifications
  • Capacity:
    100 ml
  • Seq:
    01
  • Pk:
    1
  • Side necks:
    24/40 NS
  • Centre neck:
    24/40 NS
Specifications
About this item
Borosilicate glass 3.3.
  • Heavy wall
  • Very good chemical resistance
  • Very good temperature resistance
These flasks have standard taper outer joints on the both center neck and angled side neck.