Order Entry
China
ContactUsLinkComponent
VWR® PESTINORM®, Butyl Acetate, Secondary Reference Standard for GC
VWR® PESTINORM®, Butyl Acetate, Secondary Reference Standard for GC
Catalog # VWRC85655.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® PESTINORM®, Butyl Acetate, Secondary Reference Standard for GC
Catalog # VWRC85655.180
Supplier:  VWR Chemicals
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • CAS No.:
    123-86-4
  • Description:
    Butyl acetate standard for GC
  • Description:
    Butyl acetate

Specifications

About this item

A secondary GC solvent reference standard for various chromatographic and analytical applications.

  • Purity and the specific impurity profile is determined by GC-MS and reported in the CoA
  • Traceability data to specific current lot of primary standard
  • Completely synthetic and typically over 99,9% pure by GC-FID