订单输入
China
ContactUsLinkComponent
 
VWR® PESTINORM®, Dichloromethane (DCM) Stabilised, Secondary Reference Standard for GC
目录编号 VWRC85658.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® PESTINORM®, Dichloromethane (DCM) Stabilised, Secondary Reference Standard for GC
目录编号 VWRC85658.180
Supplier:  VWR Chemicals
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • 包装类型:
    Glass bottle
  • 包装规格:
    100 mL
  • 说明:
    Dichloromethane (DCM) standard for GC

Specifications

About this item

A secondary GC solvent reference standard for various chromatographic and analytical applications.

  • Purity and the specific impurity profile is determined by GC-MS and reported in the CoA
  • Traceability data to specific current lot of primary standard
  • Completely synthetic and typically over 99,9% pure by GC-FID

Stabilised with amylene.