Order Entry
China
ContactUsLinkComponent
Anti-CD73 Rat Monoclonal Antibody (PE (Phycoerythrin)-Cyanine7) [clone: eBioTY/11.8 (TY/11.8)]
Anti-CD73 Rat Monoclonal Antibody (PE (Phycoerythrin)-Cyanine7) [clone: eBioTY/11.8 (TY/11.8)]
Catalog # EBIO25-0731-82
Supplier:  EBIOSCIENCE
Anti-CD73 Rat Monoclonal Antibody (PE (Phycoerythrin)-Cyanine7) [clone: eBioTY/11.8 (TY/11.8)]
Catalog # EBIO25-0731-82
Supplier:  EBIOSCIENCE

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    CD73
  • Clonality:
    Monoclonal
  • Clone:
    eBioTY/11.8 (TY/11.8)
  • Flow cytometry:
    Yes
  • Host:
    Rat
  • Isotype:
    IgG1
  • Reactivity:
    Mouse
  • Environmentally Preferable:
  • Form:
    Liquid
  • Gene ID:
    23959
  • Antigen synonyms:
    ecto-5'-nucleotidase|5'-NT|Purine 5-Prime-Nucleotidase|eN|E5NT|NTE|NT5|5'-nucleotidase|eNT|NT|CD73
  • Storage buffer:
    PBS, 0.1% gelatin, 0.09% sodium azide, pH 7.2
  • Storage temperature:
    4° C, store in dark, DO NOT FREEZE!
  • Concentration:
    0.2 mg/mL
  • Purification:
    Affinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Description: eBioTY/11.8 recognizes the with CD73 a 69-kDa GPI-anchored cell-surface protein with ecto-5'-nucleotidase activity. Expression on myeloid cells (CD11b) is restricted to the bone marrow. In human CD73 can be induced to secrete a soluble form with IL-2 suggesting a role in mediating activation signals. Differences between human and mouse CD73 have been reported. BALB/c mice have more CD4+CD73+ than CD8+CD73+ while the reciprocal is documented in humans. CD73 is expressed on a subset of lymphocytes and increases during lymphocyte maturation. Recently it has been found that memory CD4 T cells express and are similar to the uncommitted primed precursor helper cells (Thpp) that can differentiate into Th1 or Th2 cells. Furthermore CD73 has been found on regulatory T cells. Applications Reported: This eBioTY/11.8 (TY/11.8) antibody has been reported for use in flow cytometric analysis. Applications Tested: This eBioTY/11.8 (TY/11.8) antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.125 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Light sensitivity: This tandem dye is sensitive photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 µL cell sample + 100 µL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 488-561 nm; Emission: 775 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser Ecto-5-prime-nucleotidase (5-prime-ribonucleotide phosphohydrolase) catalyzes the conversion at neutral pH of purine 5-prime mononucleotides to nucleosides, the preferred substrate being AMP. The enzyme consists of a dimer of 2 identical 70 kD subunits bound externally to the plasma membrane by a glycosyl phosphatidyl inositol linkage. The enzyme is used as a marker of lymphocyte differentiation. Consequently, a deficiency of NT5E occurs in a variety of immunodeficiency diseases. Other forms of 5-prime nucleotidase exist in the cytoplasm and lysosomes and can be distinguished from ecto-NT5 by their substrate affinities, requirement for divalent magnesium ion, activation by ATP, and inhibition by inorganic phosphate. It is not known whether the different enzymes are coded by different genes or result from different posttranslational modifications of a single coding sequence.

Flow Cytometry: 0.125 µg/test

Type: Primary
Antigen: CD73
Clonality: Monoclonal
Clone: eBioTY/11.8 (TY/11.8)
Conjugation: PE (Phycoerythrin)-Cyanine7
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse