Anti-RORC Rat Antibody [clone: AFKJS-9]
Catalog # EBIO14-6988-82
Supplier: EBIOSCIENCE
Specifications
- Antibody type:Primary
- Antigen name:Retinoid related orphan receptor gamma
- Antigen symbol:ROR gamma (t)
- Clonality:Monoclonal
- Clone:AFKJS-9
- Conjugation:Unconjugated
- Host:Rat
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Rhesus monkey,Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:19885, 6097
- Antigen synonyms:RZRG|MGC129539|NR1F3|RORG|RZR-GAMMA|TOR
- Storage buffer:PBS, 0.09% sodium azide, pH 7.2
- Storage temperature:4° C
- Concentration:0.5 mg/mL
- Purification:Affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type:
Antigen: RORC
Clonality:
Clone: AFKJS-9
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human