Anti-RORC Rat Antibody [clone: AFKJS-9]
EBIO12-6988-82
- Antibody type:Primary
- Antigen name:RAR Related Orphan Receptor Gamma
- Antigen symbol:ROR gamma (t)
- Clonality:Monoclonal
- Clone:AFKJS-9
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG2a
- Reactivity:Rhesus monkey,Human,Mouse
- Form:Liquid
- Gene ID:19885, 6097
- Antigen synonyms:RZRG|MGC129539|NR1F3|RORG|RZR-GAMMA|TOR
- Storage buffer:PBS, 0.1% gelatin, 0.09% sodium azide, pH 7.2
- Storage temperature:4° C, store in dark, DO NOT FREEZE!
- Concentration:0.2 mg/mL
- Purification:Affinity chromatography
- Size:100 µg
- Pk:100 µG
Type:
Antigen: RORC
Clonality:
Clone: AFKJS-9
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human