Order Entry
China
ContactUsLinkComponent
Anti-TBX21 Mouse Antibody [clone: eBio4B10 (4B10, 4-B10)]
Anti-TBX21 Mouse Antibody [clone: eBio4B10 (4B10, 4-B10)]
Catalog # EBIO12-5825-82
Supplier:  EBIOSCIENCE
Anti-TBX21 Mouse Antibody [clone: eBio4B10 (4B10, 4-B10)]
Catalog # EBIO12-5825-82
Supplier:  EBIOSCIENCE

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    T-box 21
  • Antigen symbol:
    T-bet
  • Clonality:
    Monoclonal
  • Clone:
    eBio4B10 (4B10)
  • Conjugation:
    PE (Phycoerythrin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Rhesus monkey,
    Human,
    Mouse
  • Form:
    Liquid
  • Gene ID:
    57765, 30009
  • Antigen synonyms:
    T-box transcription factor TBX21|transcription factor TBLYM|T-cell-specific T-box transcription factor T-bet|T-box expressed in T cells|T-PET|T-box protein 21|T-bet|TBLYM|TBET
  • Storage buffer:
    PBS, 0.1% gelatin, 0.09% sodium azide, pH 7.2
  • Storage temperature:
    4° C, store in dark, DO NOT FREEZE!
  • Concentration:
    0.2 mg/mL
  • Purification:
    Affinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type:
Antigen: TBX21
Clonality:
Clone: eBio4B10 (4B10, 4-B10)
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, Rhesus