Order Entry
China
ContactUsLinkComponent
Anti-CD69 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: FN50]
Anti-CD69 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: FN50]
Catalog # EBIO12-0699-42
Supplier:  EBIOSCIENCE
Anti-CD69 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: FN50]
Catalog # EBIO12-0699-42
Supplier:  EBIOSCIENCE
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD69 molecule
  • Antigen symbol:
    CD69
  • Clonality:
    Monoclonal
  • Clone:
    FN50
  • Conjugation:
    PE (Phycoerythrin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Environmentally Preferable:
  • Form:
    Liquid
  • Gene ID:
    969
  • Antigen synonyms:
    EA1|MLR-3|member C|AIM|GP32/28|CLEC2C|CD69 antigen|C-type lectin domain family 2|BL-AC/P26
  • Storage buffer:
    PBS, 0.2% BSA, 0.1% gelatin, 0.09% sodium azide, pH 7.2
  • Storage temperature:
    4° C, store in dark, DO NOT FREEZE!
  • Concentration:
    5 µl/Test
  • Purification:
    Affinity chromatography
  • Size:
    100 Tests
  • Pk:
    100 Tests

Specifications

About this item

Description: The FN50 monoclonal antibody reacts with human CD69, also known as very early activation antigen (VEA). CD69 is approximately 30 kDa and is expressed on the cell-surface as a disulfide-linked dimer. CD69 is rapidly upregulated upon activation and expressed on lymphocytes, monocytes and platelets. Applications Reported: The FN50 antibody has been reported for use in flow cytometric analysis. Applications Tested: This FN50 antibody has been pre-titrated and tested by flow cytometric analysis of stimulated normal human peripheral blood cells. This can be used at 5 µL (0.015 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser Also called AIM or activation inducer molecule, CD69 is a gp28/34 disulfide bonded homodimer with a Mr of 60 kDa under non-reducing conditions. CD69 contains one or two N linked oligosacaride. The CD69 molecule is also present on activated platelets. In normal peripheral blood a variable percentage of cells express the CD69 antigen. It is involved in lymphocyte signal transduction.

Flow Cytometry: 5 µL (0.015 µg)/test

Type: Primary
Antigen: CD69
Clonality: Monoclonal
Clone: FN50
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human