Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:CD69 molecule
- Antigen symbol:CD69
- Clonality:Monoclonal
- Clone:FN50
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:969
- Antigen synonyms:EA1|MLR-3|member C|AIM|GP32/28|CLEC2C|CD69 antigen|C-type lectin domain family 2|BL-AC/P26
- Storage buffer:PBS, 0.2% BSA, 0.1% gelatin, 0.09% sodium azide, pH 7.2
- Storage temperature:4° C, store in dark, DO NOT FREEZE!
- Concentration:5 µl/Test
- Purification:Affinity chromatography
- Size:100 Tests
- Pk:100 Tests
Specifications
About this item
Description: The FN50 monoclonal antibody reacts with human CD69, also known as very early activation antigen (VEA). CD69 is approximately 30 kDa and is expressed on the cell-surface as a disulfide-linked dimer. CD69 is rapidly upregulated upon activation and expressed on lymphocytes, monocytes and platelets. Applications Reported: The FN50 antibody has been reported for use in flow cytometric analysis. Applications Tested: This FN50 antibody has been pre-titrated and tested by flow cytometric analysis of PHA-stimulated normal human peripheral blood cells. This can be used at 5 µL (0.125 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser Also called AIM or activation inducer molecule, CD69 is a gp28/34 disulfide bonded homodimer with a Mr of 60 kDa under non-reducing conditions. CD69 contains one or two N linked oligosacaride. The CD69 molecule is also present on activated platelets. In normal peripheral blood a variable percentage of cells express the CD69 antigen. It is involved in lymphocyte signal transduction.
Flow Cytometry: 5 µL (0.125 µg)/test
Type: Primary
Antigen: CD69
Clonality: Monoclonal
Clone: FN50
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human