


Methanol ≥99.9%, HiPerSolv CHROMANORM®, ULTRA for LC-MS, suitable for UPLC/UHPLC-MS instruments
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:67-56-1
:
Formula:CH₃OH
MDL Number:MFCD00004595
:32.04 g/mol
Merck Index:13,05984
Boiling Pt:64.6 °C (1013 hPa)
Melting Pt:–98 °C
Density:0.7918 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-659-6
UN:1230
ADR:3 (6.1),II
:Filtered through a 0.2 µm filter, packaged under nitrogen.
Flash Pt:11 °C (closed cup)
REACH:01-2119433307-44
:GHS02
:GHS06
:GHS08
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
Specifications
Specification Test Results
- Assay (GC):Min. 99.9 %
- Acidity:Max. 0.0002 meq/g
- Alkalinity:Max. 0.0002 meq/g
- Boiling point:64 - 65 °C
- Colouration:Max. 10 APHA
- Density (20/20):0.791 - 0.793
- Evaporation residue:Max. 1 ppm
- Water:Max. 0.02 %
- Al (Aluminium):Max. 20 ppb
- Ag (Silver):Max. 100 ppb
- Ba (Barium):Max. 100 ppb
- Ca (Calcium):Max. 200 ppb
- Cd (Cadmium):Max. 20 ppb
- Co (Cobalt):Max. 20 ppb
- Cu (Copper):Max. 10 ppb
- Fe (Iron):Max. 50 ppb
- K (Potassium):Max. 50 ppb
- Mg (Magnesium):Max. 50 ppb
- Mn (Manganese):Max. 100 ppb
- Mo (Molybdenum):Max. 20 ppb
- Na (Sodium):Max. 200 ppb
- Ni (Nickel):Max. 100 ppb
- Pb (Lead):Max. 20 ppb
- Zn (Zinc):Max. 100 ppb
- Gradient (220 nm):Max. 3 mAU
- Gradient (235 nm):Max. 2 mAU
- Gradient (254 nm):Max. 1 mAU
- Transmittance (210 nm):Min. 45 %
- Transmittance (220 nm):Min. 65 %
- Transmittance (230 nm):Min. 85 %
- Transmittance (235 nm):Min. 90 %
- Transmittance (240 nm):Min. 95 %
- Transmittance (250 nm):Min. 95 %
- Transmittance (260 nm):Min. 98 %
- Transmittance (280-400 nm):Min. 98 %
- Absorbance (210 nm):Max. 0.347
- Absorbance (220 nm):Max. 0.188
- Absorbance (225 nm):Max. 0.155
- Absorbance (230 nm):Max. 0.071
- Absorbance (240 nm):Max. 0.023
- Absorbance (250 nm):Max. 0.023
- Absorbance (260 nm):Max. 0.009
- Absorbance (280-400 nm):Max. 0.009
- Fluorescence (as quinine; 254 nm):Max. 1 ppb
- Fluorescence (as quinine) (365 nm):Max. 0.5 ppb
- Conforms Ph.Eur. R1 1053201:Passes test
- Conforms Ph.Eur. R2 1053202:Passes test
- Conforms to ACS:Passes test
- Filtered at 0.2 µm:Confirmed
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_VWRC85800.290 spec_VWRC85800.290 spec_VWRC85800.320 spec_VWRC85800.320 spec_VWRC85800.320
Product specification
VWR® LC-MS Solvents, Additives, Mixes and Associated Products