Order Entry
China
ContactUsLinkComponent
Gemini® C18 Reversed Phase HPLC Columns
Gemini® C18 Reversed Phase HPLC Columns
Supplier:  PHENOMENEX
Certificates
Gemini® C18 Reversed Phase HPLC Columns
Supplier:  PHENOMENEX
Specifications
  • Catalog No:
  • PHEN00G-4435-E0
  • PHEN00F-4439-Y0
  • PHEN00F-4435-E0
  • Carbon load:
  • 14%
  • 14%
  • 14%
  • Endcapped:
  • Yes
  • Yes
  • Yes
  • pH range:
  • 1,0 - 12,0
  • 1,0 - 12,0
  • 1,0 - 12,0
  • Surface area:
  • 375 m²/g
  • 375 m²/g
  • 375 m²/g
  • USP code:
  • L1
  • L1
  • L1
Specifications