Gemini® NX-C18 Reversed Phase HPLC Columns
Catalog # PHEN00F-4453-Y0
Supplier: PHENOMENEX
Specifications
- Column Format:Analytical
- Ø int.:3,00 mm
- Particle Size:3 µm
- Pore size:110 Å
- Length:150 mm
- Seq:02
- Endcapped:Yes
- USP code:L1
- pH range:1,0 - 12,0
- Surface area:375 m²/g
- Carbon load:14%
- Phase:NX-C18
- Pk:1
Specifications
About this item
These rugged reversed phase HPLC columns offer extended lifetime under extreme pH conditions and excellent stability for reproducible, high efficiency separations.
- Take full advantage of high and low pH conditions (pH 1 to 12) to manipulate selectivity
- Expect longer column lifetime with patented TWIN-NX™ technology
- Recommended for high efficiency analytical or preparative separations under extreme pH conditions
- Control selectivity of ionisable compounds for optimised methods
- High and low pH methods
- Consistent performance in both volatile and non-volatile buffers
TWIN-NX™ technology uses an improved patented organo-silica grafting process which incorporates highly stabilising ethane crosslinking.