Order Entry
China
ContactUsLinkComponent
VWR® Talon® Support Stands, Heavy Duty
VWR® Talon® Support Stands, Heavy Duty
Catalog # VWRU60100-105
undefined
VWR® Talon® Support Stands, Heavy Duty
Catalog # VWRU60100-105
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Support stand, with base (432×330 mm) and rod (16×457 mm)
  • Seq:
    01
  • Pk:
    1
Specifications
About this item
Multi-purpose support was originally designed to permit cylindrical vessels up to 305mm (12") in diameter to sit close to the upright rod.
  • Versatile
  • Heavy-duty design
  • Multi-purpose application
  • Supplied with stainless steel rod
  • Finished in black crackle lacquer
Support is ideal for mounting stirring motors or for heavy equipment. Each of the four corners rests on a rubber cushion. Supplied with stainless steel rod 457mm (18") high, 16mm (5/8") diameter.