Order Entry
China
ContactUsLinkComponent
Ammonium tetrathiomolybdate 99.95% (trace metals basis)
Irritant
Ammonium tetrathiomolybdate 99.95% (trace metals basis)
Certificates
undefined
Irritant
Ammonium tetrathiomolybdate 99.95% (trace metals basis)

Specification Test Results

  • Appearance:
    Brown to black Powder or crystals
  • Total trace metal impurities:
    ≤500 ppm
  • Molybdenum (Mo):
    35.5 to 38.1 %

Specification Test Results

Related Information