Isopropylmagnesium chloride - LiCl complex 1.3 M in THF, AcroSeal®
:745038-86-2
:807329-97-1
:C₃H₇ClMg·LiCl
:MFCD07784514
:145.24 g/mol
:0.98 g/cm³
:–21 °C
:3399
:4.3,I
:GHS05
:GHS02
:GHS07
:GHS08
:Ambient
- Appearance:Clear yellow to grey to brown Solution (May develop some turbidity or precipitate)
- Titration Grignard:1.1 to 1.5 M