Order Entry
China
ContactUsLinkComponent
 
 
 
Ammonium thioglycolate 70% (w/w) in water
 
undefined
 
 
Ammonium thioglycolate 70% (w/w) in water

 

  • Appearance:
    Clear colorless Solution
  • Titration Iodimetric:
    68.0 to 72.0 %
  • Identification:
    ammonium:positive
  • pH:
    5.8 to 6.4
  • Iron (Fe):
    ≤0.2 ppm