




Isobutyllithium 1.7 M in heptane, AcroSeal®
Catalog # ACRO377591000
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle, AcroSeal®
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Appearance:Colorless to yellow Solution (May develop some turbidity or precipitate)
- Titration Active base:15.0 to 17.0 %
Specification Test Results
About this item
:
:64.056 g/mol
Density:0.7 g/cm³
UN:3394
ADR:4.2,
:GHS08
:GHS05
:GHS09
:GHS02
:GHS07
Storage Temperature:Freezer
CAS Number:920-36-5
Formula:C₄H₉Li
MDL Number:MFCD01863085