Order Entry
China
ContactUsLinkComponent
Methyl-β-cyclodextrin (mixture of several Methylated), M.W. ∼ 1310
Methyl-β-cyclodextrin (mixture of several Methylated), M.W. ∼ 1310
Catalog # ACRO377110250
CAS Number:  
undefined
Methyl-β-cyclodextrin (mixture of several Methylated), M.W. ∼ 1310
Catalog # ACRO377110250
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White Powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤5.0 %
  • Degree of substitution:
    1.7 to 1.9 (NMR)
  • Specific optical rotation:
    +154.0° to +165.0° (20°C, 589 nm) (c=5, water)

Specification Test Results

About this item

 :
 :1303.3000 g/mol
Melting Pt:165…172 °C
Flash Pt:187 °C
Storage Temperature:Ambient
CAS Number:128446-36-6
MDL Number:MFCD00074980