
3-Formylsalicylic acid hydrate 97%
Catalog # ACRO377070010
Supplier: Thermo Fisher Scientific
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Yellow to beige to light brown Crystalline powder or needles
- Infrared spectrum:Conforms
- Loss on drying:≤11 % (1 g, 105°C, 3 hrs)
- HPLC:≥96.0 %
- Titration with NaOH:≥96.0 % (On dry substance)
Specification Test Results
About this item
CAS Number:137660-88-9
:
Formula:C₈H₈O₅
MDL Number:MFCD03804454
:184.146 g/mol
Melting Pt:249…252 °C
:GHS07
Storage Temperature:Ambient
CAS Number:1266615-78-4