Order Entry
China
ContactUsLinkComponent
Sodium 30% (w/w) dispersion in toluene, Ø <0.1 mm

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mL
  • Seq:
    2590
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Dispersion in toluene
  • Additional info:
    Trace analysis based on 100% sodium metal purity
  • Strength:
    ≥30 % (Na)
  • Chloride (Cl):
    ≤50 ppm
  • Calcium (Ca):
    ≤550 ppm
  • Potassium (K):
    ≤300 ppm

Specification Test Results

About this item

 :
 :22.99 g/mol
Density:0.90 g/cm³
Flash Pt:4 °C
Storage Temperature:Ambient
UN:3482
ADR:4.3,I
 :GHS08
 :GHS05
 :GHS02
CAS Number:7440-23-5
Formula:Na
MDL Number:MFCD00085307