β-Sitosterol with ca. 10% campesterol, ca. 75% beta-sitosterol
:83-46-5
:
:C₂₉H₅₀O
:MFCD00003631
:414.72 g/mol
:139…142 °C
:>100 °C
:Refrigerator
- Appearance:White Powder
- Infrared spectrum:Conforms
- Loss on drying:≤3.5 %
- GC:≥95 % Total sterols
- Specific optical rotation:-24° to -32° (20°C, 589 nm) (c=2, CHCl3)