Microtiter tubes
Microtiter tubes, PP, 1,2 ml, Natural, Bulk
Catalog # PORE845
Supplier: POREX BIOPRODUCTS
Jump to:
Product Details & Documents
For harvesting cells, cell uptakes and receptor binding.
Ideal for a wide variety of laboratory procedures, such as HTLV-III testing, RIA, EIA, PCR, pharmaceutical quality
control, blood bank sample freezer storage, transport of specimens and reagents.
规格
- Packed:Bulk
- Capacity:1,2 ml
- Colour:Natural
- Description:Microtiter tubes, PP