关于此项目
An open-platform ice-free cooler that accommodates most CoolRack® and CoolSink® modules.
- Consists of a base, cooling core and a lid
- Includes a 1 to 8 °C temperature indicator for visual assurance of proper temperature
- To use with most Corning CoolRack® and CoolSink® Modules
- The internal cooling or freezing cores provide 0,5 to 4 °C for up to 4 hours
- One module capacity
- Purple color
Low profile and small footprint make it ideal for use in the hood, keeping samples cold (0,5 to 4,0 °C) up to four hours. 1 to 8 °C temperature indicator provides visual assurance of temperature performance. To extend the cooling duration, keep an additional XT cooling core in the freezer and rotate the cores as needed.