Order Entry
China
Orders LinkContactUsLinkComponent
 

Microplate reader, Luminoskan™ Ascent

Luminoskan Ascent 100 - 240V, 50/60 Hz, with three dispensers
  HERA5300173
Microplate reader, Luminoskan™ Ascent
  HERA5300173
Microplate reader, Luminoskan™ Ascent
Microplate reader, Luminoskan™ Ascent

Luminoskan Ascent 100 - 240V, 50/60 Hz, with three dispensers

 
 
   
1 * 1 件
 

The Luminoskan Ascent is a robust, compact, microplate luminometer for luminometric research applications, such as reporter gene, immuno and cell based assays, enzyme studies, molecular interactions, nucleic acid quantification and microbiological assays.

  • Excellent optical performance
  • Versatile plate formats offered
  • Fast reading speeds
  • Up to three dispensers for reagent addition
  • Designed for robot compatibility

 : PC is not included with the instruments, but PC software, Thermo Scientific Ascent Software, is included.

 
  • Description:
    Luminoskan Ascent 100 - 240V, 50/60 Hz, with three dispensers
  • Incubation range:
    From ambient ±3...45 °C, at ambient 25 °C
  • Dispensing speed:
    25 s, 96-well plate, 5 μl/well
  • Shaking:
    Orbital shaker
  • Measurement time (96-well plate):
    15 s
  • Dynamic range:
    >9 decades over whole gain setting
  • Dispensers:
    Up to 3
  • Weight:
    Basic unit 21 kg; 3 optional dispensers - each add 3,5 kg to the weight
  • Interfaces:
    Computer control via serial RS-232C port
  • Cat. No.:
    HERA5300173
  • Dispensing volume range:
    1 - 1000 µl in 1 µl increments
  • Sensitivity:
    10 amol ATP/well using flash reaction, white 384-well plate
  • Detector:
    Photomultiplier tube
  • Wavelength selection:
    Filters
  • Model:
    Luminoskan™ Ascent
  • Wavelength range:
    270 - 670 nm
  • W×D×H:
    420×420×340 mm
  • Plate types:
    1 - 384-well plates
  • Measuring modes:
    Luminescence