Order Entry
China
ContactUsLinkComponent
Toluene 99.5% ACS for spectroscopy
Irritant
Health
Flammable
Toluene 99.5% ACS for spectroscopy
Catalog # ACRO421175000
CAS Number:  
undefined
Irritant
Health
Flammable
Toluene 99.5% ACS for spectroscopy
Catalog # ACRO421175000
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Water:
    ≤0.03 % (Coulometric)
  • GC:
    ≥99.5 %
  • UV:
    at 286 nm A: ≤1
  • Color scale:
    ≤10 APHA
  • Residue after evaporation:
    ≤0.001 %
  • Substance darkened by H2SO4:
    passes test
  • Sulfur compounds (as S):
    ≤0.003 %

Specification Test Results

About this item

CAS Number:108-88-3
 :
Formula:C₆H₅CH₃
MDL Number:MFCD00008512
 :92.14 g/mol
Boiling Pt:110.6 °C (1013 hPa)
Melting Pt:–95 °C
Density:0.866 g/cm³
Flash Pt:4 °C
UN:1294
ADR:3,II
 :GHS07
 :GHS08
 :GHS02
Storage Temperature:Ambient