

Diisopropyl ether 99+% stabilised, extra pure
:108-20-3
:
:(CH₃)₂CHOCH(CH₃)₂
:MFCD00008880
:102.18 g/mol
:68.5 °C (1013 hPa)
:–85.5 °C
:0.72 g/cm³
:–29 °C
:Ambient
:3,II
:Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
:1159
:GHS07
:GHS02
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.3670 to 1.3690 (20°C, 589 nm)
- Water:≤0.1 %
- GC:≥99.0 %
- Color scale:≤10 APHA
- Stabilizer:5 to 250 ppm BHT
- Residue after evaporation:≤0.01 %
- Acidity:≤0.005 %
- Peroxides:≤0.002 % (as H2O2)