Order Entry
China
ContactUsLinkComponent
Tris(dibenzylideneacetone)dipalladium(0), complex with chloroform 97%
Irritant
Health
Tris(dibenzylideneacetone)dipalladium(0), complex with chloroform 97%
Catalog # ACRO369340010
CAS Number:  
undefined
Irritant
Health
Tris(dibenzylideneacetone)dipalladium(0), complex with chloroform 97%
Catalog # ACRO369340010
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Purple to black Crystalline powder
  • Assay:
    ≥96.0 % (ex Pd)

Specification Test Results

About this item

 :
 :1035.11 g/mol
Melting Pt:131…135 °C
 :GHS07
 :GHS08
Storage Temperature:Freezer
CAS Number:52522-40-4
Formula:C₅₁H₄₂O₃Pd₂·CHCl₃
MDL Number:MFCD00075479