Order Entry
China
ContactUsLinkComponent
N-Methoxy-N-methylacetamide 98%
Flammable
N-Methoxy-N-methylacetamide 98%
Catalog # ACRO369310050
CAS Number:  
undefined
Flammable
N-Methoxy-N-methylacetamide 98%
Catalog # ACRO369310050
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4240 to 1.4260 (20°C, 589 nm)
  • GC:
    ≥97.5 %

Specification Test Results

About this item

 :
 :103.12 g/mol
Boiling Pt:152 °C (760 mmHg)
Density:0.97 g/cm³
Flash Pt:49 °C
Storage Temperature:Ambient
UN:1993
ADR:3,III
 :GHS02
CAS Number:78191-00-1
Formula:C₄H₉NO₂
MDL Number:MFCD00060098