



Bis(2-methoxyethyl)aminosulphur trifluoride 50% (w/w) 2.7 M in toluene
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:202289-38-1
:
Formula:C₆H₁₄F₃NO₂S
MDL Number:MFCD01321415
:221.24 g/mol
Density:1.2 g/cm³
Flash Pt:4 °C
UN:3286
ADR:3,II
:GHS06
:GHS08
:GHS05
:GHS02
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Dark yellow to orange Liquid
- NMR:≥95 % (on neat substance)
- Titration with HClO4:≥50 %