Order Entry
China
ContactUsLinkComponent
Hexachloroplatinic(IV) acid hydrate (38 - 40% Pt) 99.9% (trace metals basis)
Corrosive
Toxic
Health
Environment
Hexachloroplatinic(IV) acid hydrate (38 - 40% Pt) 99.9% (trace metals basis)
Catalog # ACRO363590010
CAS Number:  
undefined
Corrosive
Toxic
Health
Environment
Hexachloroplatinic(IV) acid hydrate (38 - 40% Pt) 99.9% (trace metals basis)
Catalog # ACRO363590010
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Orange to orange-brown Powder and/or chunks
  • Assay:
    38 to 40 % (Pt)
  • Total trace metal impurities:
    ≤0.1 %

Specification Test Results

About this item

 :
 :427.83 g/mol
Melting Pt:60 °C
Storage Temperature:Ambient
UN:2507
ADR:8,III
 :GHS05
 :GHS06
 :GHS08
 :GHS09
CAS Number:26023-84-7
Formula:H₂Cl₆Pt·H₂O
MDL Number:MFCD00149909