Order Entry
China
ContactUsLinkComponent
DL-Isocitric acid, trisodium salt hydrate 95%
  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:
  • Appearance:
    White to light yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • Water:
    ≤20 % (K.F.)
  • Titration with HClO4:
    ≥94 % (On dry substance)
 :
 :Ambient
 :1637-73-6
 :C₆H₅Na₃O₇·xH₂O
 :MFCD00149284