Order Entry
China
ContactUsLinkComponent
 
 
2-(1H-Imidazol-5-yl)acetic acid hydrochloride 99%
  ACRO204960010
 :  
undefined
 
2-(1H-Imidazol-5-yl)acetic acid hydrochloride 99%
  ACRO204960010
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

 

 

  • Appearance:
    White to light yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    221 to 225 °C
  • Titration Argentometric:
    ≥98.5 %
  • Titration with NaOH:
    ≥98.5 %

 

 

 :
 :162.58 g/mol
 :218…222 °C
 :GHS07
 :Ambient
 :3251-69-2
 :C₅H₇ClN₂O₂
 :MFCD00012698