Order Entry
China
ContactUsLinkComponent
Zinc standard solution, for AAS, 1 mg/ml Zn in 2% HCl
Zinc standard solution, for AAS, 1 mg/ml Zn in 2% HCl
Catalog # ACRO196465000
CAS Number:  
Zinc standard solution, for AAS, 1 mg/ml Zn in 2% HCl
Catalog # ACRO196465000
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    2% HCl
  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Element:
    Zinc
  • Symbol:
    Zn

Specifications

About this item